missing translation for 'onlineSavingsMsg'
Learn More

Picrotoxin, Tocris Bioscience™

Product Code. 30001203
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

GABAA antagonist

GABAA receptor antagonist; potent CNS stimulant.

Specifications

CAS 124-87-8
Molecular Formula C30H34O13
Quantity 1 g
Synonym picrotoxin
InChI Key VJKUPQSHOVKBCO-DMRKGALJSA-N
SMILES CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O
Molecular Weight (g/mol) 602.589
PubChem CID 131846146
Formula Weight 602.59
Percent Purity 97%
Chemical Name or Material Picrotoxin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.