missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 0.1% Trifluoroacetic Acid (v/v) in Water, LC-MS Grade

An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

105.00 € - 340.00 €

Specifications

Grade LC-MS
Item Name Trifluoroacetic Acid (TFA)
Content And Storage Store in a cool, dry, and well-ventilated area. Protect from direct sunlight.
Form Liquid
Reagent Type Solvent Mixture, Ion Pairing Reagent
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13414279
View Documents
Thermo Scientific™
85172
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13434279
View Documents
Thermo Scientific™
85173
4 x 1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Thermo Scientific™ 0.1% Trifluoroacetic Acid in Water is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

This 0.1% Trifluoroacetic Acid in Water is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography-and spectrometry-grade 0.1% TFA in water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Validated

    – tested more than 20 ways to ensure maximal LC/MS sensitivity

  • Purified

    – processed, filtered and sealed to extend LC/MS column life

  • Convenient

    – packaged to eliminate variability and reduce handling

Specifications

Specifications

LC-MS
Store in a cool, dry, and well-ventilated area. Protect from direct sunlight.
Solvent Mixture, Ion Pairing Reagent
HPLC, Mass Spectrometry
Trifluoroacetic Acid (TFA)
Liquid
Pierce™
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.