Learn More
Thermo Scientific™ Pierce™ 1-Step Transfer Buffer
Perform rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
40.72 € - 432.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15175204
|
Thermo Scientific™
84731 |
1 L |
111.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15185204
|
Thermo Scientific™
84742 |
200 mL | N/A | N/A | N/A | |||||
15225346
|
Thermo Scientific™
84731X5 |
5 x 1 L |
432.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
The Thermo Scientific™ Pierce 1-Step Transfer Buffer is designed for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
When used with the Pierce G2 Fast Blotter, this buffer provides effective gel-to-membrane transfer of proteins in 5 to 10 minutes with efficiency that is equal to, or better than, conventional Western blotting techniques. Pierce 1-Step Transfer Buffer is also compatible with other protein semi-dry transfer devices that are paired with a suitable high-current power supply. Such devices provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.
Highlights:
- Fast – high ionic strength formulation provides 5- to 10-minute protein transfer when used with compatible fast semi-dry blotting systems
- Ready to use – supplied as a 1X solution that is stable and ready to use at room temperature
- Optimized – designed for seamless and reliable performance with the Pierce G2 Fast Blotter
- Compatible – effective with other semi-dry transfer devices equipped with a suitable high-current power supply
- Economical – excellent performance without the special or costly consumables that are required by other fast semi-dry transfer devices
Required: Pierce G2 Fast Blotter (Part No. 62288) or other semi-dry transfer device that is equipped or paired with a capable high-current power supply.
Specifications
Transfer Buffer |