missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plate Seal for Pierce™ 96-well Microdialysis Plates

Product Code. 12311983 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 Sheet
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12311983

Brand: Thermo Scientific™ 88269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Simultaneously process 1 to 96 samples with 10 to 100 μL volumes with this convenient and disposable rapid dialysis system that is automation compatible.

The Thermo Scientific™ Pierce 96-well Microdialysis Plate, 10K MWCO is a convenient and disposable rapid dialysis system for the simultaneous processing of 1 to 96 samples with volumes 10 to 100 μL and is compatible with automation systems.

Thermo Scientific Pierce Plate Seal for Pierce 96-well Microdialysis Plates is an adhesive coated plastic sheet, perforated for use with an 8-well strip microdialysis plate. Sufficient For: 12 x 8 microdialysis plate wells.

These plate seals are made of high-quality polypropylene and help protect samples from contamination and evaporation, especially during long-term storage.

TRUSTED_SUSTAINABILITY

Specifications

Sufficient For 12 x 8 microdialysis plate wells
Content And Storage Upon receipt store at room temperature. Product shipped at ambient temperature.
Format Microdialysis Plates
Product Line Slide-A-Lyzer
Quantity 1 Sheet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.