missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Albumin Serum Depletion Kits

Product Code. 13484309 Shop All Thermo Scientific Products
Click to view available options
Description:
Albumin Depletion Kit
Albumin and IgG Removal Kit
Unit Size:
1 set
24 reactions
This item is not returnable. View return policy

Product Code. 13484309

Brand: Thermo Scientific™ 85160

Please to purchase this item. Need a web account? Register with us today!

Edge
Este artículo no se puede devolver. Vea la política de devoluciones

Optimize serum sample prep by depleting unwanted albumin and IgG serum proteins for protein-profiling methods using quality depletion kits.

Concentrations of albumin and IgG can make up more than 70% of total serum proteins and often mask lower quantity proteins during protein-profiling experiments. Optimize your serum sample prep by removing albumin or IgG quickly and conveniently. By using our depletion kits to remove excess high-abundance serum proteins, less abundant proteins of interest can be easily detected by 2D electrophoresis and other protein profiling methods such as mass spectrometry.

Pierce Albumin/IgG Removal Kit

Optimized—microcentrifuge spin cup method is scaled for treating 10 μL human serum samples for 2D electrophoresis
Efficient—removes nearly all HSA and IgG, enabling low-abundance proteins to be detected in 2D gels
Fast—processes samples in less than 40 minutes
Scalable—use different amounts of affinity resin to match sample volume and concentration

The Thermo Scientific Pierce Albumin/IgG Removal Kit uses a classical mixture of Cibacron Blue Dye and Protein A resin for removal of albumin and IgG. The dye binds to albumin with good specificity, and Protein A binds to many species and subclasses of IgG. The Pierce Albumin/IgG Removal Kit is economical and can be used to treat samples from other species besides human (e.g., monkey, swine, rabbit). The kit is also scalable and sufficient to process 25 samples, each containing 600 μg of total serum protein (about 10 μL of serum).

Pierce Albumin Depletion Kit

Complete—includes optimized buffer and microcentrifuge spin columns to remove albumin quickly and conveniently from 10- to 50-μL samples
Adaptable—easy-to-dispense slurry improves performance, eases processing of multiple samples, and can be adapted to larger or smaller columns or 96-well filter plates
Compatible—removing excess albumin facilitates MS analysis or electrophoresis gel analysis of low-abundance serum proteins

The Thermo Scientific Pierce Albumin Depletion Kit resin is supplied as a 50% slurry. To prepare the depletion columns, 200 μL of slurry is dispensed into the supplied microcentrifuge spin columns to obtain 100 μL of settled beads to process 10 to 50 μL of serum sample in a single reaction. The kit has been optimized to bind human, porcine, and sheep albumin from serum samples. With a modification to the protocol, albumin from bovine, calf, goat, and rat can be removed with this method. The kit is not effective for removal of mouse albumin. Processed samples are ready for immediate downstream analysis by electrophoresis or MS applications.

TRUSTED_SUSTAINABILITY

Especificaciones

Description Albumin Depletion Kit
For Use With (Equipment) Microcentrifuge, Mass Spectrometer
Product Type Albumin Depletion Kit
Product Line Pierce
Sufficient For Removing albumin from 24 human serum samples, 10 to 50 μL each
Content And Storage • Pierce Albumin Depletion Resin, 5 mL
• Binding/Wash Buffer, 11 mL
• Pierce Spin Columns, 24 columns

Store at 4°C.
Starting Material Plasma, Serum
Final Product Type Protein
Detection Method Mass Spectrometry
Format Kit
Workflow Step Sample Enrichment, Sample Clean Up
Quantity 24 Reactions
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Pierce Albumin Serum Depletion Kits
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
Indicaciones de peligro
  • H319-Causes serious eye irritation.
Consejos de prudencia
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : Sodium chloride
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado