Learn More
Thermo Scientific™ L-Lysine-2HCl, 4,4,5,5-D4 for SILAC
Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.
170.00 € - 1028.00 €
Specifications
For Use With (Equipment) | Mass Spectrometer |
---|---|
Product Type | Amino Acids |
Content And Storage | Store at room temperature. |
Label or Dye | Mass Isotope of Amino Acid |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11305402
|
Thermo Scientific™
88437 |
50 mg |
170.00 €
50mg |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
12106490
|
Thermo Scientific™
88438 |
500 mg |
1028.00 €
500mg |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
Description
Thermo Scientific™ Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.
SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of “heavy“ 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.Highlights:
Efficient – 100% label incorporation into proteins of living cells
Flexible – different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases
Multiplex capabilities – several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment
High-quality supplements – heavy amino acids with >99% isotope purity
Specifications
Mass Spectrometer | |
Store at room temperature. | |
Room Temperature | |
Protein | |
Protein Labeling |
Amino Acids | |
Mass Isotope of Amino Acid | |
Cells | |
Mass Spectrometry | |
Solid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.