missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Anti-DYKDDDDK Magnetic Agarose

Thermo Scientific™ Pierce™ Anti-DYKDDDK Magnetic Agarose Beads provide a fast, convenient method for purification of DYKDDDK-tagged recombinant proteins

Brand:  Thermo Scientific™ A36798

1501.00 EUR valid until 2025-12-31
Use promo code "24353" to get your promotional price.


 View more versions of this product

Product Code. 15883521

  • 1662.00 € / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce Anti-DYKDDDDK Magnetic Agarose provides a fast, convenient method for purification and immunoprecipitation (IP) of DYKDDDDK-tagged proteins from in vitro protein expression systems, bacteria, yeast, and mammalian cells. The amino acid sequence DYKDDDDK, commonly known as 'FLAG', is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to a magnetite-embedded agarose core particle.

For protein purification, the magnetic agarose is added to a sample containing DYKDDDDK-tagged proteins with the tag on either the N- or the C-terminus. Captured proteins are then magnetically separated from the supernatant, and non-specifically bound proteins can be washed away before dissociating bound DYKDDDDK-tagged proteins with elution buffer. The magnetic agarose is removed from the solution using a magnetic stand or an instrument such as the KingFisher Flex Magnetic Particle Processor. Automated instruments are especially useful for higher throughput purifications and screening of purification conditions.

Features include:

  • Specific—unique base beads and highly specific antibody minimizes off-target binding (low non-specific binding)
  • High purity—optimized bind-wash-elute protocol enables high purity
  • High yield—special antibody conjugation method enables high yield
  • Rapid—entire purification protocol typically takes less than 40 mins
  • Economical—purification protocol allows multiple reuses
  • Versatile—beads are compatible with manual and automated workflows (e.g., KingFisher instruments)

Characteristics of Pierce Anti-DYKDDDDK Magnetic Agarose:

  • Composition: anti-DYKDDDDK antibody covalently attached to a magnetic, highly crosslinked agarose support
  • Magnetization: ferrimagnetic with low remanence
  • Bead size: 10–40 μm
  • Bead concentration: 25% slurry in phosphate buffered saline, 0.01% Tween-20 detergent, 0.02% sodium azide, pH 7.2
  • Binding capacity: ≥3.2 mg DYKDDDDK-tGFP-His protein (∼32 kDa)/mL settled beads
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pierce™
5 mL
25 to 30 μm
5 mL settled beads. 20 mL supplied as a 25% suspension in PBS containing 0.02% sodium azide.

Store at 2–8°C.
Liquid
Wet Ice
Magnetic Agarose
Magnetic Agarose
20 mL
Slurry: 25%
20 mL
Anti-DYKDDDDK monoclonal antibodies
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI399898 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberA36798
Lot NumberZG399860 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberA36798
Lot NumberZF399859 Certificate TypeCertificate of Analysis Date19/06/2024 Catalog NumberA36798
Lot NumberZC387751 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberA36798
Lot NumberZC379077 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog NumberA36798
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Anti-DYKDDDDK Magnetic Agarose > 20 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.