Learn More
Thermo Scientific™ Pierce™ Avidin
Perform Western blot, ELISA or other biotin-based detection methods using these purified and HRP-, AP- or FITC-labeled avidin molecules.
Brand: Thermo Scientific™ 21128
154.67 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Thermo Scientific™ Pierce Avidin and Conjugates include purified avidin protein from hen egg whites, plus avidin-HRP, avidin-AP and other conjugated forms for use in biotin-based detection methods.
Avidin from chicken egg whites is a glycoprotein that effectively binds biotin, a tag that is frequently used to tag antibodies and other probes for immunodetection methods. Pure avidin is a key component in certain immunohistochemistry (IHC) methods. Alkaline phosphatase (AP) and horseradish peroxidase (HRP) conjugates of avidin are useful for Western blotting or ELISA detection with appropriate enzyme substrates.Highlights:
Native avidin – glycoprotein from chicken egg whites, 67kDa, pI equals 10
Soluble – glycosylation and isoelectric point renders avidin more soluble than streptavidin
Affordable – significantly less expensive than recombinant streptavidin
ABC staining – avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
Pure protein and conjugates – choose pure, unconjugated avidin for use in ABC staining or peroxidase or alkaline phosphatase forms for membrane and plate-based assays
Specifications
Unconjugated | |
Store at -20°C. Shipped at ambient temperature. | |
Pierce™ | |
Biotin |
20 mg | |
Solid | |
Avidin (Unconjugated) | |
Unlabeled |
For Research Use Only. Not for use in diagnostic procedures.