missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ BCA Protein Assay Reagent A
Estimate visually, or measure with a standard spectrophotometer or plate reader at 562 nm.
109.00 € - 668.00 €
Specifications
Content And Storage | Store at room temperature. |
---|---|
Description | Pierce BCA Protein Assay Reagent A |
Assay | BCA Assay |
Detection Method | Colorimetric |
Product Line | Pierce™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10752735
|
Thermo Scientific™
23222 |
3.75 L |
668.00 €
3.75 L |
In Stock
Log in to see stock. |
|||||
10198543
|
Thermo Scientific™
23221 |
250 mL |
109.00 €
250mL |
In Stock
Log in to see stock. |
|||||
11881345
|
Thermo Scientific™
23223 |
1 L |
240.00 €
1000mL |
In Stock
Log in to see stock. |
|||||
10056623
|
Thermo Scientific™
23228 |
500 mL |
135.00 €
500mL |
In Stock
Log in to see stock. |
|||||
Description
Thermo Scientific™ Pierce BCA Protein Assay Kit remains one of the most popular protein quantitation methods worldwide.
Additional Information and Online Ordering
- A component of the Pierce BCA Protein Assay Kit
- Kit is a two-component, high-precision, detergent-compatible assay that is used for total protein concentration determination compared to a protein standard
- Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research
- The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications
- Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy
- Estimate visually or measure with a standard spectrophotometer or plate reader at 562 nm
- Exhibits less protein-to-protein variation than dye-binding methods (Bradford)
- Unaffected by typical concentrations of most ionic and non-ionic detergents
- Assay has a 30 min. incubation; much easier and four times faster than classical Lowry methods
- Linear working range for BSA of 20 to 2000 μg/mL
- Detect down to 5 μg/mL with the enhanced protocol
- Store at room temperature
Specifications
Store at room temperature. | |
BCA Assay | |
Pierce™ | |
Not Target-Specific | |
Spectrophotometer, Microplate Reader |
Pierce BCA Protein Assay Reagent A | |
Colorimetric | |
Protein Quantitation Assay | |
Solution-based Detection, Absorbance |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title