missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Biotin Quantitation Kit

HABA dye reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in antibodies and other proteins.

Brand:  Thermo Scientific™ 28005

Product Code. 10341634

  • 307.00 € / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Biotin Quantitation Kit includes aliquots of HABA reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in labeled antibodies and other proteins.

Determine the molar ratio of biotin incorporated into a protein using the HABA-avidin method. The HABA dye (4'-hydroxyazobenzene-2-carboxylic acid) binds to avidin to produce a yellow-orange colored complex that absorbs at 500nm. Free biotin present in solution with this avidin-HABA complex will displace the HABA dye and cause the absorbance to decrease.The amount of biotin present can be calculated directly from the decreased absorbance at 500nm. The Thermo Scientific EZ Biotin Quantitation Kit contains pre-measured doses of the Avidin-HABA mixture and biotinylated horseradish peroxidase positive control to simplify reagent preparation and minimize the amount of waste generated. The convenient assay can be performed either in a cuvette or in a microplate.

Highlights:

  • HABA-avidin complex can be used over a wide range of pH and salt concentrations
  • Amount of biotin can be calculated directly from the decreased absorbance at 500nm complexing with the HABA dye

Eliminate the need for cumbersome assay equations with a convenient HABA calculator found at www.piercenet.com/HABA/habacalc.cfm

Includes:

HABA/Avidin Premix, 24 tubes; Biotinylated HRP, 12 tubes

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Biotin Quantitation Kit
Sufficient For: Testing biotinylation of 12 protein samples
• HABA-Avidin Premix, 24 tubes
• Biotinylated Horseradish Peroxidase, 5 mg

Store at 4°C.
Biotin
12 Tests
Pierce™
Antibodies, Proteins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.