missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Biotinylated Protein Interaction Pull-Down Kit

Easily capture and affinity-purify binding partners of any biotin-labeled protein or other ligand using these streptavidin agarose beads and complete reagent set.

Brand:  Thermo Scientific™ 21115

Product Code. 10035833

  • 654.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Beschreibung

Beschreibung

Thermo Scientific™ Pierce Biotinylated Protein Interaction Pull-Down Kit contains the necessary components to capture and purify interactors of a biotin-labeled protein or ligand.

The kit makes it possible to perform an efficient pull-down assay with high protein recovery and reproducibility. This protocol uses immobilized streptavidin to capture a biotinylated protein and a spin column format that enables complete gel retention during each capture, wash and elution step. When the biotinylated bait protein is immobilized, it can withstand stringent washes and remains bound during prey protein elution, which results in an isolated prey protein without bait protein contamination. This technique is ideal when using native or recombinant proteins that are not expressed with a fusion tag. Additionally, prey protein is isolated from the immobilized bait protein using native binding conditions and nondenaturing elution conditions. The Biotinylated Protein Interaction Pull-Down Kit is designed to teach the method to first-time users and to increase ease-of-use and convenience for experienced researchers.

Highlights:

  • Biotin pull-down – purifies protein interactors of any biotinylated protein or biotin-tagged ligand

  • Complete kit – provides all components and detailed protocol for purifying protein:protein interactions

  • No special equipment needed – uses common laboratory equipment and reagents (e.g., microcentrifuge)

  • Spin column efficiency – microcentrifuge spin columns facilitate microcentrifuge spin columns facilitate simple and efficient manipulation of streptavidin agarose beads, including simple processing of multiple samples

Includes:

Streptavidin agarose resin, TBS pack, biotin blocking buffer, wash buffer, elution buffer, spin columns and collection tubes

Recommended for:

Discover a new protein interaction complex using a biotinylated protein as the bait to capture a putative binding partner from a cell lysate or other sample; Confirm expression of a known protein interaction target in a cell lysate; Determine the ability of two purified proteins to interact (bind) in various buffer conditions; Extract protein:protein interaction information from in vitro transcription/ translation lysates

TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

Sufficient For: At least 25 pull-down reactions, each using 25 μL of resin
• Streptavidin Agarose Resin, 1.5 mL
• TBS Pack (makes 500mL), 1 pack
• Biotin Blocking Solution, 15 mL
• Wash Buffer (pH 5.0), 100 mL
• Elution Buffer (pH 2.8), 50 mL
• Pierce Spin Columns and Accessories, 27 columns
• Microcentrifuge Collection Tubes, 200 tubes

Store at 4°C
Microcentrifuge
25 reactions kit
Not Target-Specific
25 Reactions
Affinity Purification
Pierce™
Pull-Down (Biotin-Based)
Pull-Down Kit
Kit
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Sonderangebote

Sonderangebote

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt

For Research Use Only. Not for use in diagnostic procedures.