missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate)

Crosslink amines to amines with these long (21.7A and 35.8A) crosslinkers composed of NHS-ester groups and 5-unit or 9-unit PEG spacer arms that enhance protein solubility.

Brand:  Thermo Scientific™ 21581

 View more versions of this product

Product Code. 11821245

  • 380.00 € / 100mg
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce BS(PEG)n Crosslinkers include BS(PEG)5 and BS(PEG)9, which are bis-succinimide ester-activated PEG compounds for crosslinking between primary amines in proteins and other molecules.

BS(PEG)5 is a bis-succinimide ester-activated PEG compound used for crosslinking between primary amines (—NH2) in proteins and other molecules. The N-hydroxysuccinimide ester (NHS) groups at either end of the PEG5 spacer react specifically and efficiently with lysine and N-terminal amino groups at pH 7-9 to form stable amide bonds.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

Features of BS(PEG)5:

  • Reactive groups: NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • NHS esters react with—NH2 groups at pH 7-9, forming stable amide bonds
  • Water-soluble; compare to non-pegylated BS3 (also soluble) (Cat. No. 21580) and DSS (water-insoluble) (Cat. No. 21655)
  • Membrane-impermeable, allowing for cell surface labeling
  • Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
  • Polyethylene glycol spacer arms help maintain conjugate solubility
  • Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Ideal for small molecule or peptide conjugations

Alternative Names for BS(PEG)n:

  • Bis-NHS-dPEG
  • NHS-PEG-NHS
  • BS(PEO)5
  • Bis(NHS)PEO5
  • BS-PEO9
  • dPEG NHS ester
  • Bis-succinimide ester PEG 400

Product References

  • Crosslinker Application Guide -- search for recent literature references for this product
  • Bioconjugate Techniques, Academic Press (Cat. No. 20036)

Related Products

  • BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate) (Cat. No. 21582)
  • BS3 (bis(sulfosuccinimidyl)suberate) (Cat. No. 21580)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amine-Amine
BS(PEG)5
Yes
Upon receipt store desiccated at -20°C.
Ambient
Chemical Labeling
NHS Ester
Liquid
Water
Yes
No
532.5
21.7 Å
No
Pierce™
Homobifunctional
Medium (10 to 30 Å)
100 mg
Standard, Single-use
Crosslinker
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.