Learn More
Thermo Scientific™ BS2G-d0 (bis(sulfosuccinimidyl) glutarate-d0)
Crosslink amines to amines using these heavy (deuterated) and light matched pairs of a 7.7A, NHS-ester crosslinker for mass spectrometry analysis of protein interactions.
Brand: Thermo Scientific™ 21610
Description
Thermo Scientific™ Pierce BS2G-d0/d4 reagents form a matched set of heavy (deuterated) and light mass varieties of a short, amine-reactive crosslinker that facilitates the identification of crosslinked peptides by mass spectrometry.
Highlights:
- Well-characterized, high-purity, deuterium-labeled crosslinker and its hydrogen-containing analog
- Instructions edited by an expert in the crosslinking/MS strategy field
- Suberate- and glutarate-based reagent pair - provides a “molecular ruler” option to study inter- and intra-molecular distances to gain structural information not possible with just one reagent
- Efficient and convenient - requires only microgram amounts of protein; an excellent alternative to NMR or X-ray crystallography-based methods that require large amounts of protein, special solvents or crystal formation
- Reactive groups: Sulfo-NHS ester (homobifunctional)
- Reactive toward: amino groups

Specifications
Amine-Amine | |
BS2G-d0 | |
No | |
Upon receipt store desiccated at 4 to 8°C. | |
Ambient | |
Chemical Labeling | |
Sulfo-NHS Ester | |
Powder | |
Water | |
Yes |
No | |
530.35 | |
7.7 Å | |
No | |
Pierce™ | |
Homobifunctional | |
Short (<10 Å) | |
10 mg | |
Standard | |
Crosslinker |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.