missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Chromatography Cartridge Accessory Pack

Accessory pack of caps and adaptors to fit 1mL and 5mL cartridges of desalting or affinity resins for manual or automated liquid chromatography equipment.

Brand:  Thermo Scientific™ 89971

Product Code. 10782335

  • 41.24 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Chromatography Cartridges are convenient, reliable, ready-to-use, pre-packed 1mL and 5mL columns of sample-prep and affinity-purification resins for manual or automated liquid chromatography.

The cartridge-fittings are compatible with the popular automated liquid-chromatography systems or for manual syringe processing. The cartridges attach directly to AKTA™ or FPLC Systems without additional connectors. Cartridges can be used individually or connected in a series to obtain even higher column capacity. Each product supplied in the Pierce Chromatography Cartridge format includes an accessory pack that readily adapts cartridges for use with Luer-Lok™ Syringe Fittings or tubing. The cartridges provide fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.

Highlights:

  • Two sizes– 1mL and 5mL, convenient for typical research scales

  • Compatible– fittings allow connection with popular LC systems or a standard syringe

  • Versatile– use singly or connected in series to service different capacity requirements

  • Validated– available cartridges have been tested to ensure performance in the format

  • Reusable– accessory pack includes caps for convenient storage between uses

  • Economical– comparable performance at lower cost than other commercially available cartridges

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chromatography Cartridge Accessory
Pierce™
Sufficient For: Attaching and replacing fittings for products supplied in Pierce Chromatography Cartridges
• Bottom plug, 1
• Cap, 1
• Luer-Lok Adapter, 1
• Tube Fitting, 1
1 pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.