missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Cypridina Luciferase Glow Assay Kit

Obtain bright and stable bioluminescence signal (half-life greater than 1 hour) in the presence of Cypridina luciferase reporters.

140.00 € - 780.00 €

Specifications

Type Assay Kit
Target Luciferase, Cypridina Luciferase
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12201622
View Documents
Thermo Scientific™
16170
100 Reaction Kit
140.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12201632
View Documents
Thermo Scientific™
16171
1000 Reaction Kit
780.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Thermo Scientific™ Pierce Cypridina Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life greater than 1 hour) in the presence of Cypridina luciferase reporters.

This Pierce Luciferase Glow Assay Kit contains reagents for measuring Cypridina luciferase activity in mammalian cell culture media and lysates. When used with Thermo Scientific Cypridina Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for secreted or intracellular detection of luciferase activity. The light output generated by the luciferase reaction can be correlated to the amount of luciferase protein produced, which in turn is proportional to the activity of the promoter driving the luciferase expression. The kit can be used to detect the activity of any Cypridina luciferases or derivatives that use Vargulin as a substrate.

Highlights:

  • Sensitive – highly sensitive detection of Cypridina luciferase activity

  • Secreted luciferase – allows real-time assays and kinetic studies without destroying cells

  • Stable – reagents provide increased signal stability compared to flash assays

  • Simple – does not require a luminometer with injectors

  • Compatible – assay reagents compatible with other Cypridina or Vargula luciferases

  • Automation-friendly – amenable to high throughput assays

  • Convenient – contains a universal cell lysis buffer and optimized glow assay reagent

  • Safe – allows one to perform non-radioactive assays

Includes:

Cell lysis buffer, reaction buffer and substrate

Requires:

Cypridina luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers

Recommended for:

Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Secretory pathway / protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies

Specifications

Specifications

Assay Kit
Reporter Enzyme, Luciferase Reporter Assay
Bioluminescence
384-well plate, 96-well plate
Chemical Substrate
Luciferase Substrate
Vargulin (Cypridina Luciferin)
Luciferase, Cypridina Luciferase
Mammalian Cells
Luminometer (Microplate)
Enzyme Labeled
Enhanced Chemiluminescence
Pierce™
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce Cypridina Luciferase Glow Assay Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methanol

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.