Learn More
Thermo Scientific™ Pierce™ Dilution-Free™ Tubestrip Holder
The Pierce Dilution-Free Tubestrip Holder is a reusable foam stand for use with the Pierce Dilution-Free BSA Protein Standards. Protein standard concentrations are printed on the Tubestrip Holder for easy reference and pipetting on the benchtop.
Brand: Thermo Scientific™ A57184
9.00 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
The Pierce Dilution-Free Tubestrip Holder is a reusable foam stand for use with the Pierce Dilution-Free BSA Protein Standards. Protein standard concentrations are printed on the Tubestrip Holder for easy reference and pipetting on the benchtop.
Pierce Dilution-Free BSA Protein Standards
Pierce Dilution-Free BSA Protein Standards are high-quality bovine serum albumin (BSA) reference samples for generating accurate standard curves and calibration controls in total protein assays. They are packaged as a set of seven pre-diluted standards in a multichannel pipette-friendly tubestrip, eliminating the need for serial dilutions. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.
Specifications
Pierce™ Dilution-Free™ | |
Tubestrip holder | |
1 each |
For Research Use Only.