missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Dimethylsulfoxide (DMSO), LC-MS Grade

A sequencing-grade preparation with high purity and consistent absorptivity properties, for use in HPLC and mass spectrometry methods.

Brand:  Thermo Scientific™ 85190

67.24 EUR valid until 2025-02-13
Use promo code "24126" to get your promotional price.


Product Code. 13494279

  • 75.00 € / 50mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Dimethylsulfoxide (DMSO) is a sequencing-grade preparation with high purity and consistent absorptivity properties, for use in HPLC and mass spectrometry methods.

A sequencing-grade preparation with high purity for use in traditional amino acid analyzers and other high-volume applications.

This sequencing-grade dimethylsulfoxide is >99.5% pure DMSO. The economically priced 950mL bottles are an excellent choice for protein and peptide applications that requiring large volumes of the water-miscible solvent. The 50mL bottles are provide convenience and minimize waste for small dissolution and reaction volumes. The purity test ensures that preparation is essentially devoid of amines. The formulation was originally developed for peptide sequencing applications with amino acid analyzers, which required high purity solvent and an affordable price. This DMSO is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • High-purity (low amine) solvent for various protein, peptide and amino acid applications
  • Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps
  • Suitable for HPLC detection, although it is not specifically tested for low UV-absorptivity

Product Details:

  • General properties of DMSO
  • Alternative names: Methylsulfinylmethane, methyl sulfoxide
  • Molecular formula: C2H6SO
  • Molecular weight: 78.13
  • Density: 1.101 g/mL
  • CAS number: 67-68-5

Pierce sequencing-grade DMSO is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

LC-MS
50 mL
Liquid
Pierce™
Amber glass bottle
Dimethylsulfoxide (DMSO)
Store at room temperature.
Solvent (Pure)
Liquid Chromatography, Mass Spectrometry
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.