Learn More
Thermo Scientific™ DMP (dimethyl pimelimidate)
Crosslink amines to amines with dimethyl pimelimidate (CAS 58537-94-3); composed of imidoester groups and a membrane-permeable, 7-atom spacer arm.
Brand:  Thermo Scientific™ 21666
 View more versions of this product
Product Code. 10097093
  • 63.00 € / 50mg
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Pierce DMP is dimethyl pimelimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of a 7-atom spacer arm.
Highlights:
  • Water-soluble
  • Rapid reaction with amines at alkaline pH values (pH 8-10)
  • Amidine bond retains net charge character of protein
  • Reversible at high pH values
  • Reactive groups: imidoester (homobifunctional)
  • Reactive toward: amino groups
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amine-Amine
DMP
No
Upon receipt store desiccated at 4°C.
Ambient
Chemical Labeling
Imidoester
Powder
Water
Yes
No
259.17
9.2 Å
Yes
Pierce™
Homobifunctional
Short (<10 Å)
50 mg
Standard
Crosslinker
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • DMP (dimethyl pimelimidate)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : dimethyl heptane-1,7-diimidate dihydrochloride