missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ DSP, No-Weigh™ Format
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 1 mg
1 g
Unit Size:
1g
50mg
Pack of 10
Beschreibung
Thermo Scientific™ Pierce DSP Crosslinker (Lomant's Reagent) is an amine-reactive crosslinker with NHS-ester reactive ends and a cleavable disulfide bond in its spacer arm.
Highlights:
- Cleavable by reducing agents
- Membrane-permeable
- Acylate in aqueous or organic media within two minutes at room temperature
- Four- to five-hour half-life of ester at pH 7.0
- Reactive groups: NHS ester (homobifunctional)
- Reactive toward: amino groups
Spezifikation
Spezifikation
| Chemical Reactivity | Amine-Amine |
| Cleavable | By Thiols |
| Description | DSP |
| Molecular Weight (g/mol) | 404.42 |
| PEGylated | No |
| Spacer Arm Length | 12.0 Å |
| Content And Storage | Upon receipt store desiccated at 4°C. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- DSP (dithiobis(succinimidyl propionate)), Lomant's Reagent
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : disuccinimido dithiobispropionate
For Research Use Only. Not for use in diagnostic procedures.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur