missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL

Product Code. 17175762 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2 Plates
2 x 2 Plates
2 x 25 Columns
25 Columns
5 Columns
5 x 5 Columns
Format:
96-well Filter Plate
Spin Column
Volume (Metric):
0.5 mL
10 mL
2 mL
5 mL
96-well
Unit Size:
Each
Pack of 2
Pack of 25
Pack of 4
Pack of 5
Pack of 50
This item is not returnable. View return policy

Product Code. 17175762

Brand: Thermo Scientific™ A44296S

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Enable fast and efficient removal of non-reacted fluorescent dyes, biotin, reducing agents, and crosslinkers from protein samples with Thermo Scientific Pierce Zeba Dye and Biotin Removal columns and filter plates.

Quickly remove non-reacted fluorescent dyes, biotin, reducing agents, and crosslinkers from protein samples with a 7K MWCO in fewer than 15 minutes with Thermo Scientific Pierce Zeba Dye and Biotin Removal columns and filter plates.

Removing free-dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. Pierce Zeba Dye and Biotin Removal resin is highly specialized to produce exceptional protein recoveries while effectively removing non-conjugated fluorescent dyes, biotinylation reagents, reducing agents, and crosslinkers. When using the appropriate amount of sample and working within buffer conditions, these columns and filter plates can successfully remove almost any fluorescent dye. Even dilute protein samples (25 μg/mL) can be successfully processed to remove dyes, biotin, reducing agents, and crosslinkers and to efficiently recover proteins (>7 kDa). These products are not recommended for desalting or buffer exchange.

Features of Zeba Dye and Biotin Removal columns and filter plates include:
Versatile—removes unreacted fluorescent dyes, biotinylation reagents, crosslinkers, and reducing agents from protein solutions
High recovery—low-binding resin maximizes protein recovery
Easy-to-use—no cumbersome column preparation or equilibration
Fast—small-molecule removal and protein recovery in less than 15 minutes
Flexible—available in spin columns and filter spin plates for a range of needs

Sample capacities
• 0.5-mL spin column: 50 to 120 μL sample
• 2-mL spin column: 400 to 700 μL sample
• 5-mL spin column: 1 to 2 mL sample
• 96-well filter plate: 40 to 100 μL sample (per well)

Available formats
Spin columns: 0.5, 2, 5, and 10 mL
96-well filter plates: pre-dispensed; compatible with centrifugation for manual or automated purification; enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput clean-ups

Applications
• Ideal for post-reaction clean-up of samples
• To effectively remove non-conjugated dye, biotin, reducing agents, and crosslinkers with exceptional protein recovery

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Format Spin Column
Column Type Proprietary Resin
Product Line Zeba
Quantity 5 Columns
Volume (Metric) 0.5 mL
Sample Volume (Metric) 50 to 120 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.