missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ ECL Plus Western Blotting Substrate

Acridan-based chemiluminescent and chemifluorescent HRP substrate for Western blot detection using X-ray film or CCD- or laser-based imagers.

153.00 € - 838.00 €

Specifications

Product Type Substrate
Detection Method Chemiluminescence, Fluorescence
Form Liquid
Membrane Compatibility Nitrocellulose, PVDF
Product Line Pierce™
View More Specs

Products 3
Product Code Brand Quantity Sufficient For Price Quantity & Availability  
Product Code Brand Quantity Sufficient For Price Quantity & Availability  
11547910
View Documents
Thermo Scientific™
32134
25 mL Kit 250 cm2 of membrane
153.00 €
25mL
Please to purchase this item. Need a web account? Register with us today!
11527271
View Documents
Thermo Scientific™
32132X3
300 mL Kit 3000 cm2 of membrane
838.00 €
Pack of 3
Please to purchase this item. Need a web account? Register with us today!
11517271
View Documents
Thermo Scientific™
32132
100 mL Kit 10 mini-gel blots, 1000 cm2 of membrane
350.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific™ Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent substrate to detect peroxidase activity from HRP-conjugated probes by Western blotting using X-ray film, CCD or laser-based imagers.

Thermo Scientific Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for western blot detection using X-ray film or CCD- or laser-based imagers. This product is sold as Pierce ECL 2 Substrate (Cat. #PI80196X3) through Fisher Scientific and other channels.

Features of ECL Plus Substrate:

  • Easy to use—can be substituted for the discontinued GE ECL Plus Substrate without any re-optimization
  • Higher sensitivity—detect targets down to the low-picogram level
  • Longer signal duration—sustained light output for as long as 5 hours
  • More imaging options—X-ray, CCD or laser-based imagers
  • More affordable—high quality and performance at a lower price than other suppliers' similar substrates

Pierce ECL Plus Substrate generates acridinium esters when it reacts with HRP. As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers. Pierce ECL Plus Substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. Its broad dynamic range, high sensitivity and long-lasting signal make Pierce ECL Plus Substrate an excellent choice for western blot analysis.

Specifications

Specifications

Substrate
Liquid
Pierce™
Chemiluminescence, Fluorescence
Nitrocellulose, PVDF
Low picogram to mid-femtogram
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce ECL Plus Western Blotting Substrate

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Ethanol

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ ECL Plus Western Blotting Substrate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.