missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ ECL Western Blotting Substrate

Value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.

Brand:  Thermo Scientific™ 32106

220.67 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 10005943

  • 331.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce ECL Western Blotting Substrate is a value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.

Pierce ECL Western Blotting Substrate provides reliability and performance equivalent to other standard enhanced chemilumescent (ECL) substrates for detection of horseradish peroxidase (HRP) enzyme activity, yet costs much less. Because the luminol and peroxide reagent-formulations are identical to other commercially available substrate products, one can switch to Pierce ECL without needing to optimize probing conditions or incubation protocols. Simply switch to Pierce ECL and save a bundle.

Highlights:

  • ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes

  • About half the cost of other ECL Substrates – low overhead and a commitment to customer value enables Thermo Fisher Scientific to make this product available to you at a significantly lower price than other companies (these claims are based on the 2010 U.S. list prices)

  • No optimization required – switching to Pierce ECL substrate from other entry-level ECL substrates does not require optimization or protocol changes

  • A name you can can rely on – count on the strong quality assurance, technical support and reputation behind Thermo Scientific Pierce Protein Research Products

Includes:

Detection Reagent 1 and 2

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pierce ECL Western Blotting Substrate
Chemiluminescence
Western Blot
Pierce™
HRP
2°: 1:1-15K (0.07-1μg/mL), 1°: 1:1K (0.2- 10μg/mL)
1 to 2 hr.
Luminol/Enhancer, 250mL, Stable Peroxide Buffer, 250mL
Substrate
Liquid
Nitrocellulose, PVDF
500 mL
Horseradish Peroxidase
Low Picogram
5000 sq. cm
Upon receipt store at 2-8°C. Product is shipped at ambient temperature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK400255 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number32106_LITL18 PROMO,32106
Lot NumberZK400255A Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number32106_LITL18 PROMO,32106
Lot NumberZI397794 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number32106_LITL18 PROMO,32106
Lot NumberZH395613A Certificate TypeCertificate of Analysis Date12/09/2024 Catalog Number32106_LITL18 PROMO,32106
Lot NumberZH395613 Certificate TypeCertificate of Analysis Date12/09/2024 Catalog Number32106_LITL18 PROMO,32106
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ ECL Western Blotting Substrate > 500mL kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.