Learn More
Thermo Scientific™ Ethidium Bromide Solution (10 mg/mL)
Visualize DNA fluorescently in a variety of molecular biology applications using these dilute (0.625mg/mL) or concentrated (10mg/mL) ethidium bromide solutions.
Brand: Thermo Scientific™ 17898
187.34 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.
Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.
Highlights:
0.625mg/mL EtBr Solution:
- Each drop contains 25μg of ethidium bromide and is ideal for a 50mL agarose gel
- After adding one drop of solution per 50mL of gel, the final concentration of EtBr will be 0.5μg/mL, which is the recommended concentration for electrophoresis of nucleic acids
- Can also be used as the running buffer or for staining gels after electrophoresis
10mg/mL EtBr Solution:
- A safe and convenient way to use ethidium bromide while minimizing exposure
Specifications
Store at room temperature, protected from light. | |
Fluorescence | |
Ethidium Bromide Solution | |
RNA, DNA |
In-Gel Detection | |
Ethidium bromide | |
5 x 10 mL |
For Research Use Only. Not for use in diagnostic procedures.