missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ethylene Glycol

Specially purified 50% ethylene glycol solution to provide antifreeze protection and freezer storage stability for antibody-enzyme conjugates.

Brand:  Thermo Scientific™ 29810

Product Code. 10454355

  • 160.00 € / 200mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce Ethylene Glycol Solution provides exceptional antifreeze protection and storage stability for antibody-enzyme conjugates because it is purified to remove impurities commonly found traditional glycerol stocks.

Highlights:

  • Specially purified to remove impurities such as aldehydes, peroxides, iron and UV absorbing hydrocarbons
  • Suitable for enzyme storage without the worry of losing enzymatic activity
  • Stable for months
TRUSTED_SUSTAINABILITY
Specifications

Specifications

200 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK401988 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number29810
Lot NumberZF359238 Certificate TypeCertificate of Analysis Date21/06/2024 Catalog Number29810
Lot NumberZA362570 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number29810
Lot NumberYD356648 Certificate TypeCertificate of Analysis Date20/04/2023 Catalog Number29810
Lot NumberSPECS Certificate TypeCertificate of Analysis Date16/03/2023 Catalog Number29810
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Ethylene Glycol > Ethylene glycol solution (50%); 200mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.