missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ F(ab')2 Micro Preparation Kit

Selectively digest whole human and other IgG antibodies with immobilized pepsin protease to make F(ab')2 fragments that retain antigen binding activity.

Brand:  Thermo Scientific™ 44688

Product Code. 10381214

  • 606.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce F(ab')2 Preparation Kits use immobilized pepsin protease to selectively digest whole human and other IgG antibodies to make F(ab')2 fragments that retain antigen binding activity.

These kits use pepsin, a nonspecific endopeptidase, to enzymatically digest the Fc portion of whole IgG to yield the fragment known as F(ab')2. This fragment is composed of a pair of Fab' units connected by two disulfide bonds. Because the pepsin protease is supplied in immobilized form as beaded agarose resin, the digestion reaction is easily stopped by removing the resin from the IgG solution; the result is digest products that are enzyme-free. An optimized procedure, high-performance desalting and Protein A spin columns make these kits easy to use.

Highlights:

  • Enzyme-free digestion products – immobilized pepsin (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the protease

  • Suitable for human and other species of IgG – the kit procedure is optimized for human, rabbit and mouse IgG, but pepsin-based digestion is effective for other species and subclasses of IgG (Note: for best results with mouse IgG1, use Part No. 44980)

  • High capacity – kit provides for digestion and purification from as much as 4mg of IgG at a time. Use the standard size kit for fragmenting 0.25 to 4mg IgG, and the micro kit for 25 to 250μg IgG

  • Complete and easy to use – kits include all reagents, spin columns and accessories needed to prepare and purify antibody fragments

Includes:

Pepsin agarose resin, digestion buffer, Protein A columns with binding and elution buffers, spin desalting columns and accessories

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pierce F(ab')2 Micro Preparation Kit
Pierce™
ERK5/BMK1 (pan)
10 Antibody Samples, each containing 25 to 250 μg IgG
Sufficient For: 10 antibody samples, each containing 25 to 250 μg IgG
• Pierce Pepsin Agarose, 0.5 mL
• F(ab')2 Digestion Buffer, 55 mL
• NAb Protein A Plus Spin Columns, 0.2 mL, 2 columns
• PBS Packs (each makes 500 mL), 2 packs
• IgG Elution Buffer, 50 mL
• Zeba Spin Desalting Columns, 7K MWCO, 0.5 mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK398789 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number44688
Lot NumberZK398789A Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number44688
Lot NumberZF395955 Certificate TypeCertificate of Analysis Date19/06/2024 Catalog Number44688
Lot NumberZF395955A Certificate TypeCertificate of Analysis Date19/06/2024 Catalog Number44688
Lot NumberZC394204 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number44688
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ F(ab')2 Micro Preparation Kit > Micro Kit for 25 to 250μg IgG samples; 0.5mL-resin kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.