missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluoraldehyde™ o-Phthaldialdehyde Crystals (OPA)

Measure peptides or amino acids (Ex 340nm, Em 455nm) with phthalaldehyde, a fluorescent, amine-reactive, HPLC pre- or post-column labeling reagent.

Brand:  Thermo Scientific™ 26015


Product Code. 11821405

  • 417.00 € / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Fluoraldehyde (OPA) is o-phthalaldehyde, a highly sensitive fluorescent pre- or post-column derivatization reagent for peptide or amino acid detection and quantitation in HPLC.

Fluoraldehyde Crystals are a specially purified, fluorogenic-grade preparation of o-phthalaldehyde (OPA). This amine-reactive compound is primarily used for high-sensitivity, fluorescent detection of primary amines, including amino acids, peptides, proteins and polyamines in pre- or post-column chromatographic effluents.

Highlights:

  • For pre- or post-column amino acid derivatization for fluorescent detection and quantitation

  • Reacts with all primary amine-containing analytes to yield fluorescent derivatives (ex/em = 340nm/455nm)

  • Provides an accurate measure of both composition and absolute protein-peptide content

  • Ideal for work with recombinant proteins and synthetic peptides

  • Can be used for fluorescent protein or peptide assay

  • Can be prepared as a stable aqueous solution in borate (see instructions)

Reagent Properties:

  • Can be prepared as a stable aqueous solution in borate (see instructions)

  • Alternative names: Fluoraldehyde Crystals, OPA

  • Chemical name: ortho-Phthalaldehyde

  • Reactive toward: Primary amines (–NH2)

  • Chemical formula: C8H6O2

  • CAS number: 643-79-8

  • Molecular weight: 134.12

  • Form: Pale yellow, free-flowing crystals

TRUSTED_SUSTAINABILITY
Specifications

Specifications

o-Phthaldialdehyde
Fluoraldehyde™
OPA (o-phthaldialdehyde)
5 g
Amine
Store at room temperature.
Fluorescence
Solid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • o-Phthalaldehyde

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : phthalaldehyde

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.