missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Formic Acid, LC-MS Grade

A high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.

132.00 € - 206.00 €

Specifications

Item Name Formic Acid
Form Liquid
Reagent Type Solvent (Pure)
Product Line Pierce™
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10628654
View Documents
Thermo Scientific™
28905
10 x 1 mL
132.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
13454279
View Documents
Thermo Scientific™
85178
50 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Thermo Scientific™ Formic Acid is high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.

Formic acid is a component found in reverse-phase mobile phases to provide protons for LC/MS analysis. The presence of a low concentration of formic acid in the mobile phase is also known to improve the peak shapes of the resulting separation. Unlike trifluoroacetic acid (TFA), formic acid is not an ion-pairing reagent, and it does not suppress MS ionization of polypeptides when used as a mobile phase component.

Highlights:

  • Prescored, nitrogen-flushed, amber glass to protect formic acid from light and moisture
  • 99% purity for consistent LC baselines and no interference introduced into LC and mass spectrometry applications
  • Convenient format simplifies preparation of gradient and isocratic mobile phases containing 0.1% (v/v) formic acid in water or acetonitrile
  • Contents of a single vial in a final volume of 1L solvent yields a mobile phase of the most common formic acid concentration

Ordering Alerts:

  • X in the ordering table indicates that hazardous shipping charges apply.

Storage Requirements:

  • Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Specifications

Specifications

Formic Acid
Liquid
Solvent (Pure)
Pierce™
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Formic Acid, 99+%, 10 x 1 mL Ampules

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.

Supplemental information
  • MIXTURE LIST-Contains : Formic acid

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.