missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Gaussia Luciferase Glow Assay Kit

Product Code. p-4532166 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12142240

Brand: Thermo Scientific™ 16160

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Obtain bright and stable bioluminescence signal (half-life greater than 1 hour) in the presence of Gaussia luciferase reporters

The Thermo Scientific™ Pierce Gaussia Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life greater than 1 hour), especially in the presence of Gaussia-Dura luciferase reporter.

The Thermo Scientific Pierce Gaussia Luciferase Glow Assay Kit provides a sensitive bioluminescence system to determine expression of a gene of interest with a luciferase reporter. The extremely bright and stable bioluminescence signal (half-life greater than 1 hour) is produced in the presence of the Gaussia-Dura luciferase reporter due to oxidation of the luciferase substrate coelenterazine (Figure 1). Gaussia luciferase is a 20 kDa protein from the marine copepod, Gaussia princeps, that has greater protein stability and signal brightness than firefly and native Renilla luciferase. The bioluminescent enzyme is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. Light output generated by the luciferase reaction can be correlated to the amount of Gaussia luciferase protein produced and used to determine the activity of the promoter driving Gaussia expression.

Features of the Gaussia Luciferase Glow Assay Kit:
Sensitive—highly sensitive detection of Gaussia luciferase activity
Stable—increased signal stability
Simple—does not require a luminometer with injectors
Compatible—assay reagents compatible with other Gaussia luciferases
Automation-friendly—amenable to high throughput assays
Convenient—contains a universal cell lysis buffer and optimized glow assay reagent
Safe—allows one to perform non-radioactive assays

When used with Thermo Scientific Gaussia-Dura Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for detection of secreted or intracellular luciferase activity from promoter or pathway activation with extended signal stability. The light output generated by the luciferase reaction (emission maximum of 485 nm) indicates that Gaussia luciferase was transcribed. This light emission can be correlated to the amount of luciferase protein produced, which in turn is proportional to the activity of the promoter for Gaussia luciferase expression and indicates that the regulatory region of the target gene was active. This reaction does not require adenosine triphosphate (ATP) or other cofactors. The kit includes cell lysis buffer, reaction buffer and substrate and can be used to detect the activity of any Gaussia luciferases or derivatives that use coelenterazine as a substrate.

The Gaussia Luciferase Glow Assay Kit requires:
Gaussia luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Varioskan LUX Microplate Readers.

Applications of the Gaussia Luciferase Glow Assay Kit:
• Promoter studies for analyzing cis- regulatory elements and trans-acting factors
• Drug screening
• siRNA and miRNA screening
• Multiplexed assays to study off-target effects
• Secretory pathway/protein localization reporter assays
• Signal transduction pathway analysis
• RNA splicing studies

The Gaussia-Dura gene used in the Thermo Scientific Gaussia-Dura Luc Vectors is derived from the wild type Gaussia gene and provides equally bright but more stable light output. The Gaussia Glow Assay Kit reagent yields a Gaussia-Dura luciferase reaction with glow-type kinetics that provides a lower intensity but longer-duration signal allowing for higher sample throughput. This glow-type assay with a prolonged luminescence signal does not require reagent injectors and is recommended for luminometers without injectors or for applications requiring batch processing of samples.

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Gaussia Luciferase
Sufficient For 100 Luciferase Assays
Quantity 100 Reaction Kit
Content And Storage Sufficient For: 100 luciferase assays in microplate wells
• Gaussia Glow Assay Buffer, 5 mL (store at 4°C)
• 100X Coelenterazine, 50 μL (store at -80°C)
• Luciferase Cell Lysis Buffer (2X), 6 mL (store at room temperature)

Kit may be stored at -80°C, or individually at indicated temperatures.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate Coelenterazine
Promoter EF-1α
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pierce Gaussia Luciferase Glow Assay Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.