missing translation for 'onlineSavingsMsg'
Learn More

Pierce™ Pierce™ Glutathione Magnetic Agarose Beads

Thermo Scientific™ Pierce™ Glutathione Magnetic Agarose Beads provide a fast, convenient method for purification of glutathione-S-transferase (GST) from a bacterial, yeast, or mammalian crude cell lysate

Brand:  Pierce™ 78602

 View more versions of this product

Product Code. 15585899

  • 744.00 € / 5mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Product features:
Pierce Glutathione Magnetic Agarose Beads are well suited for purifying GST fusion proteins from a soluble protein extract and are ideal for the purification of proteins expressed at low levels from diluted supernatants.

The beads can be used in manual applications with a magnetic stand or automated applications with an instrument such as the Thermo Scientific KingFisher™ Flex Magnetic Particle Processor. Automated instruments are especially useful for higher throughput purification and screening of purification conditions.
  • High-performance beads—non-aggregating, magnetite (Fe3O4), superparamagnetic beads provide exceptional uniformity for both manual and automated HTS applications
  • Stable affinity ligand—glutathione is covalently immobilized to particles, enabling leach-resistance and clean purification products
  • High capacity—binding capacity is sufficient for both routine and demanding purification procedures
Applications:
  • Protein purification and isolation
  • High throughput protein purification using automated magnetic particle processor
Target molecule: GST
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mL settled resin. Supplied as 20 mL of 25% v/v suspension in 20% ethanol.

Store at 2–8°C.
Liquid Suspension
20 mL
Glutathione
Wet Ice
Pierce™
Magnetic Agarose Bead
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.