Learn More
Thermo Scientific™ Pierce™ Glutathione Superflow Agarose
Perform high-purity, large-scale purification of GST-tagged proteins with glutathione that has been immobilized to a crosslinked 6% beaded agarose.
Brand:  Thermo Scientific™ 25239
5898.70 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 13444199
  • 9670.00 € / 1 L
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Perform high-purity, large-scale purification of GST-tagged proteins with Thermo Scientific™ Pierce Glutathione Superflow Agarose, which consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose.
Additional Information and Online Ordering
Pierce Glutathione Superflow Agarose is a highly crosslinked, durable resin that does not compress at flow rates required for medium- to large-scale FPLC purifications. The resin holds up well to a variety of chemical and pH values, and it is compatible with common clean-in-place procedures. The specific binding of glutathione to the GST-tag results in high purity and high yield in a single step purification process.
Highlights:
  • High dynamic binding capacity – binds 10mg of pure GST per mL of resin at a linear flow rate of 150cm/hour; can bind greater than 20mg/mL at 30cm/hour flow rate
  • High purity – provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates
  • Robust – highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200cm/hour
  • Compatible – maintains function after exposure to a wide variety of chemicals and pH values
  • Cost effective – resin is stable through multiple cycles of cleaning and reuse
Recommended for:
Medium- to large-scale FPLC purification of GST-tagged proteins
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pierce Glutathione Superflow Agarose
Glutathione onhighly crosslinked 6% beaded agarose (superflow 6), slurried in water with sodium azide
1 L
Binding up to 30mg GST/mL resin (static capacity) or up to 10mg GST/mL resin (dynamic capacity)