Learn More
Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL
Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.
245.00 € - 810.00 €
Specifications
Capacity (Metric) | Up to 2,000,000 EU/mL |
---|---|
Formulation | 22mL-capacity centrifuge columns containing 1mL resin |
Content And Storage | Upon receipt store at 4°C. |
Form | Liquid |
Product Line | Pierce™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12391973
|
Thermo Scientific™
88276 |
5 Columns |
245.00 €
Pack of 5 |
In Stock
Log in to see stock. |
|||||
12201203
|
Thermo Scientific™
88277 |
25 Columns |
810.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||||
Description
Reduce endotoxin levels in protein samples by greater than 99% in one hour while maximizing protein recovery with Thermo Scientific™ Pierce High Capacity Endotoxin Removal Resin.
The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(epsilon-lysine), as an affinity ligand to selectively bind endotoxins. Endotoxin levels in protein samples are reduced by greater than 99% in as little as 1 hour using our spin cup format, and protein recovery is ≥85%. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers. Pierce High capacity Endotoxin Removal Resin is available as a slurry to pack a custom column that can be re-used at least 5 times with no loss in endotoxin removal efficiency. This resin is also available in a new single-use spin column format optimized for different sample volumes.Highlights:
High capacity bind up to 2,000,000 EU/mL to eliminate at least 99% of endotoxins
Durable re-use resin up to 10 times with no loss in performance
Selective recover at least 85% of your protein sample
High performance compatible with FDA guidelines by reducing final EU concentration to less than 5 EU/mL
Fast our new spin cup format enables endotoxin depletion within 1 hour
Cleansingle-use spin columns avoid cross contamination of samples
Optimized spin cups are optimized for different sample volumes
Economical large-volume discounts available
Requires:
Endotoxin-free water
Recommended for:
Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection
Specifications
Up to 2,000,000 EU/mL | |
Upon receipt store at 4°C. | |
Pierce™ | |
Spin Column | |
Spin Column |
22mL-capacity centrifuge columns containing 1mL resin | |
Liquid | |
Endotoxin Removal | |
Proprietary Ligand | |
Affinity, Cellulose Resin |
Safety and Handling
- Pierce High Capacity Endotoxin Removal Spin Columns, 1 mL
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H351-Suspected of causing cancer.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
For Research Use Only. Not for use in diagnostic procedures.