missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Capacity Ni-IMAC MagBeads, EDTA compatible

Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads are designed for purification of over-expressed His-tagged proteins from cell culture media.

84.00 € - 4315.00 €

Specifications

Binding Capacity Up to 80 mg of 6 xHis-tagged protein / mL of settled magnetic beads
Binding Property Chelator Stable
Matrix Magnetic Agarose
pH Range 2 to 14 pH stability
Shipping Condition Wet Ice
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17134759
View Documents
Thermo Scientific™
A50588
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17113269
View Documents
Thermo Scientific™
A50588
1 mL
87.00 €
0.25mL
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
 
17123269
View Documents
Thermo Scientific™
A50589
5 mL
381.00 €
1.25mL
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
 
17133269
View Documents
Thermo Scientific™
A50590
25 mL N/A
N/A
17143269
View Documents
Thermo Scientific™
A50591
100 mL
4315.00 €
25mL
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
Description

Description

Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads have high binding capacity (up to 80 mg of protein per mL of settled beads) and can tolerate the high EDTA and reducing agent concentrations that are used to inhibit metalloproteases and stabilize sensitive intracellular proteins.

Features of the high-capacity EDTA-compatible Ni-IMAC MagBeads include:

  • Chelator stable—stable in buffer containing 20 mM EDTA and DTT
  • Compatible with purification of overexpressed, secreted proteins in cell culture media (e.g., Expi293, ExpiCHO, and ExpiSf expression systems)
  • Exceptional high protein binding capacity—up to 80 mg/mL of settled beads
  • High-performance—non-aggregating, superparamagnetic microparticles provide exceptional performance with manual and automated HTS applications (e.g., Thermo Scientific KingFisher instruments)

Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads consist of crosslinked agarose beads embedded with magnetite. The density of the ligand on the magnetic agarose bead results in a binding capacity similar to or better than traditional agarose resins with the added benefits of having magnetic properties and being EDTA compatible. The MagBeads are a valuable tool for small-scale purification of multiple His-tagged proteins and for scouting expression and purification conditions to be used in larger-scale purifications.

Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads are optimized for use with high-throughput magnetic platforms, such as the KingFisher 96 and KingFisher Flex magnetic particle processors, but the beads also compatible with simple benchtop purification applications using an appropriate magnetic stand. Automated instruments, however, are especially useful for higher throughput purification and screening of purification conditions.

Specifications

Specifications

Up to 80 mg of 6 xHis-tagged protein / mL of settled magnetic beads
Magnetic Agarose
Wet Ice
Magnetic Affinity Bead
Chelator Stable
2 to 14 pH stability
30 μm
Bottle
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.