missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Capacity Protein A MagBeads, alkali stable

The Pierce High-Capacity Protein A MagBeads, Alkali Stable are a high-performance magnetic bead for purifying antibodies from cell culture supernatant, ascites, serum, and crude extracts.

Brand:  Thermo Scientific™ A53037

1111.80 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 17884880

  • 2180.00 € / 25mL
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High-performance magnetic beads for purifying antibodies from cell culture supernatant, ascites, serum, and crude extracts
  • The ligand is produced in yeast and is immobilized on magnetic agarose with an average particle size of ∼30 mm
  • Engineered to withstand standard CIP procedures with 0.1M NaOH, enabling efficient cleaning of the beads for reuse
  • Beads can be regenerated at least 10 times without sacrificing binding capacity
  • Suitable for purifying mAbs and a subpopulation of scFv’s and Fab fragments containing a VH3 domain
  • Compatible with manual and automated workflows (e.g., KingFisher instruments)
  • High-affinity Protein A is securely immobilized to magnetic agarose to prevent leaching of ligand, target antibody, or base support
  • High binding capacity of ≥ 40 mg HIgG/mL settled beads
  • Animal component-free raw materials
TRUSTED_SUSTAINABILITY
Specifications

Specifications

≥40 mg HIgG/mL settled beads
Provided in the form of a 25% slurry.

Upon receipt store at 2–8°C.

25 mL
Alkaline Stable Protein A
High Capacity Magnetic Agarose Beads
∼ 30 μm
Antibody Purification
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.