missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Intact Protein Standard Mix
229.00 € - 792.00 €
Specifications
For Use With (Equipment) | Mass Spectrometer |
---|---|
Product Type | Intact Protein Standard Mix |
Product Line | Pierce™ |
Shipping Condition | Wet Ice |
Workflow Step | Calibration |
Description
Specifications
Mass Spectrometer | |
Pierce™ | |
Calibration |
Intact Protein Standard Mix | |
Wet Ice | |
Powder |
For research use only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title