Learn More
Thermo Scientific™ Pierce™ Iodoacetic Acid
Get rapid carboxymethylation of reduced cysteine residues with Thermo Scientific™ Pierce™ Iodoacetic Acid and Iodoacetamide.
Brand: Thermo Scientific™ 35603
Code : A0
Additional Details : Weight : 1.81440kg
Description
Get rapid carboxymethylation of reduced cysteine residues with Thermo Scientific™ Pierce™ Iodoacetic Acid and Iodoacetamide.
Iodoacetic acid reacts with sulfhydryls on cysteines, imidazolyl side chain nitrogens of histidines, the thioether of methionine and the primary amine group of lysines.The rate of reaction and specificity is dependent on the ionization level, which can be manipulated by the pH of the reaction condition.
Highlights:
- React at slightly alkaline pH for specific S-carboxymethylation of free sulfhydryls
- React at low pH for specific carboxymethylation of methionines
- React at high pH to favor carboxymethylation of histidines and lysines
- Methylate reduced cysteine peptide fragments in protease digests for mass spectrometry
Specifications
Thiol | |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. | |
Blocking Groups | |
Short (<10 Å) | |
500 mg | |
Iodoacetic Acid |
Iodoacetic Acid | |
Unreactive Group | |
Pierce™ | |
Powder | |
Water |
For Research Use Only. Not for use in diagnostic procedures.