Learn More
Thermo Scientific™ Pierce™ Luciferase Cell Lysis Buffer (2X)
Lyse adherent cells or pelleted cell suspensions for analysis of promoter expression in luciferase assays with this optimized protein extraction reagent.
Brand:  Thermo Scientific™ 16189
170.28 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 12142250
  • 258.00 € / 250mL
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Pierce Luciferase Cell Lysis Buffer effectively lyses adherent cells or pelleted suspension cells for analysis of promoter expression in luciferase assays.
Pierce Luciferase Cell Lysis Buffer is effective for preparing high-yield mammalian cell lysates from either adherent cells or pelleted suspension cells for analysis in luciferase reporter assays. The reagent is compatible with all Thermo Scientific Pierce Luciferase Assay Kits. The cell lysis buffer has been validated for use with COS-7, NIH3T3, Jurkat and other cell types. Lysates are produced rapidly and easily for luciferase assays and are also compatible with BCA and Coomassie (Bradford) protein assays.
Highlights:
  • Compatible with all Pierce Luciferase Reporter Assays
  • Enables rapid and efficient lysis
  • Tested compatibility with multiple Mammalian cell lines
  • Compatible with protein assays (e.g., Coomassie Plus and BCA)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Lysis Buffer
250 mL
Store at room temperature.