Learn More
Thermo Scientific™ EZ-Link™ Maleimide Activated Horseradish Peroxidase
Activated horseradish peroxidase (HRP) enzyme and kit to label proteins, peptides or other ligands via sulfhydryl groups (reduced cysteines) to make HRP conjugates.
Brand: Thermo Scientific™ 31485
298.30 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce Maleimide-Activated HRP Conjugation Kit can be used to directly prepare HRP conjugates from proteins, peptides or other ligands that contain sulfhydryl groups, such as reduced cysteines.
The Thermo Scientific Maleimide Activated Horseradish Peroxidase (HRP) Kit can be used to directly prepare HRP conjugates from proteins, peptides or other ligands that contain a free sulfhydryl group. This kit eliminates the first step of the two-step maleimide method, making conjugate preparation much simpler. The kit is supplied with a stable, Maleimide-activated horseradish peroxidase. HRP conjugates can be purified by gel filtration chromatography, ultrafiltration or dialysis, depending upon the size of the conjugated protein.Highlights:
- Prepare enzyme conjugates from proteins that contain a free sulfhydryl
- Includes 2-MEA to generate free sulfhydryls from disulfide bonds
- Includes SATA to add free sulfhydryls to lysine residues

Specifications
Thiol | |
Maleimide | |
Note: Total weight is ∼25mg as a result of buffer components and salts lyophilized along with the activated protein Storage: Upon receipt store product at 4°C. Product is shipped at ambient temperature. |
|
Solid | |
Enzyme Labeled |
5 mg | |
Water | |
Activated horseradish peroxidase lyophilized from PBS | |
EZ-Link™ | |
HRP (Horseradish Peroxidase) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.