Learn More
Thermo Scientific™ Pierce™ Maleimide Activated Plates
Thermo Scientific™ Pierce Maleimide Activated Plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.
216.00 € - 230.00 €
Specifications
Binding Property | Standard Capacity, Covalent |
---|---|
Content And Storage | Note: Plates are supplied pre-blocked with bovine serum albumin. Upon receipt store plates at 4°C protected from moisture. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature. |
Product Line | Pierce |
Material | Polystyrene |
Volume (Metric) Well | 100 μL |
Product Code | Brand | Description | Detection Method | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Detection Method | Price | Quantity & Availability | ||||
10328733
|
Thermo Scientific™
15150 |
Clear, 8-well Strip | Colorimetric |
216.00 €
Pack of 5 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
10348643
|
Thermo Scientific™
15152 |
White, 96-well | Chemiluminescence |
225.00 €
Pack of 5 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
10475015
|
Thermo Scientific™
15153 |
Black, 96-well | Fluorescence |
230.00 €
Pack of 5 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
Description
Pierce Maleimide Activated Plates provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production. Maleimide Activated Plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
Pierce Maleimide Activated Plates provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production. Maleimide Activated Plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
Highlights
• Spontaneous immobilization of cysteine-containin peptides and proteins with free sulfhydryls
• Pre-blocked to reduce nonspecific binding
Specifications
Standard Capacity, Covalent | |
Pierce | |
100 μL | |
Maleimide | |
Blocker™ BSA Buffer |
Note: Plates are supplied pre-blocked with bovine serum albumin. Upon receipt store plates at 4°C protected from moisture. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature. |
|
Polystyrene | |
96 | |
5 Plates |