missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MS(PEG)4 Methyl-PEG-NHS-Ester Reagent
Pegylate amines (-NH2 groups) on proteins (lysine residues) or assay surfaces using MS(PEG)n, amine-reactive (NHS ester), methyl-capped PEG compounds (4 to 24 EG units).
Marca: Thermo Scientific™ 22342
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Thermo Scientific™ Pierce MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) activated as NHS esters for covalent pegylation of primary amines on proteins (e.g., lysines) or assay surfaces.
MS(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and amine-reactive NHS-ester groups at opposite ends. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-NHS Ester, where the subscript “n” denotes 4, 8, 12, or 24 ethylene glycol units. The N-hydroxysuccinimide (NHS) ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.Highlights:
- NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
- Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
- Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Easy-to-follow instructions increase the likelihood of a successful outcome
Especificaciones
Amine | |
Polyethylene Glycol (PEG), Blocking Groups | |
Active Ester, Succinimidyl Ester, NHS Ester | |
Solid | |
Water |
Store desiccated at -20°C. | |
Pierce | |
4 ethylene glycol units | |
1 g | |
NHS-Ester Reagent |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only. Not for use in diagnostic procedures.
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido