Learn More
Thermo Scientific™ Pierce™ Micro-Spin Columns
Perform small-scale affinity methods, such as immunoprecipitation assays with agarose beads using these empty spin columns and cups (5 to 500μL).
Brand: Thermo Scientific™ 89879
97.85 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Description
Thermo Scientific™ Pierce Microcentrifuge Spin Cups and Spin Columns are convenient tools for manipulating small volumes of affinity supports for protein purification.
Additional Information and Online Ordering
These microcentrifuge spin cups and spin columns are convenient tools for manipulating small volumes of affinity supports (5 to 500μL) for immunoprecipitation (IP) and pull-down purification assays. Five styles of polypropylene columns and matching collection tubes offer a variety of options to suit particular types of experimental needs. Add the affinity resin and sample to one of the columns and use a microcentrifuge to efficiently wash away contaminants and elute your purified sample without losing any resin in the process. Spin columns allow you to affinity-purify more protein in less time.
Highlights:
- Efficient washing of samples means fewer washes are needed to remove contaminating proteins
- Efficient elution of samples means better recovery of immunoprecipitation and assay products
- Filter cups and columns eliminate contamination of eluted samples with affinity resin
- Filter cups and columns eliminate the need to decant supernatant from pelleted debris or resin
- Spin protocols drastically reduce the time required for small-scale affinity protocols
- Low protein-binding polypropylene column construction minimizes nonspecific binding
Spin Cups-Paper Filter with Collection Tubes (Part No. 69700):
- Column volume: 600μL
- Resin volume: 20 to 300μL
- Filter type: paper, 10μm pore size
- Cap type: collection tube cap fits onto inserted spin cup
Spin Cups-Cellulose Acetate Filter with Collection Tubes (Part No. 69702):
- Column volume: 800μL
- Resin volume: 20 to 400μL
- Filter type: cellulose acetate, 0.45μm pore size
- Cap type: collection tube cap fits onto inserted spin cup
- Spin ColumnsScrew Cap with Luer-Lok
Spin Columns-Screw Cap (Part No. 69705):
- Column Volume: 900μL
- Resin volume: 20 to 400μL
- Filter type: polyethylene filter, 10μm pore size; small and large frit options for different sample sizes
- Cap type: O-ring screw top caps; press-in bottom plugs
- Luer-Lok Adapters enable use with syringe-based kits
Spin Columns-Snap Cap with Collection Tubes (Part No. 69725):
- Column volume: 1000μL
- Resin volume: 20 to 500μL
- Filter type: polyethylene filter, 30μm (20 to 50μm) pore size
- Cap type: snap cap on column (no cap on collection tube); press-on bottom caps
Micro-Spin Columns (Part No. 89879):
- Column volume: 400μL
- Resin volume: 5 to 100μL
- Filter type: polyethylene filter, 30μm (20 to 50μm) pore size
- Cap type: O-ring screw top caps; press-on bottom caps
Microcentrifuge Tubes (Part No. 69715):
- 2mL capacity
- Dolphin-style bottom with attached snap cap
- Included and compatible with Part No. 69700
Microcentrifuge Tubes (Part No. 69720):
- 2mL capacity
- Short, conical bottom with attached snap cap
- Included and compatible with Part No. 69702
Specifications
2 mL | |
Store at room temperature. | |
50 columns | |
Microcentrifuge, Spin Column |
Pierce Micro-Spin Columns | |
Pierce™ | |
Spin Column | |
Empty Column, Polyethylene Filter |
For Research Use Only. Not for use in diagnostic procedures.