missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Modified Lowry Protein Assay Kit

Clear-to-purple (A750nm), 2-component, folin phenol- and copper-based reagent system to measure total protein concentration vs. protein standard.

Brand:  Thermo Scientific™ 23240

212.04 EUR valid until 2024-12-31
Use promo code "21989" to get your promotional price.



Product Code. 10677904

  • 231.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Modified Lowry Protein Assay is a classical, well-characterized copper-based method for measuring protein concentration.

Additional Information and Online Ordering

The Modified Lowry Protein Assay Kit combines a stabilized formulation of the original Lowry Reagents and the essential Folin-Ciocalteu Phenol reagent in a complete kit for accurately determining protein concentration in a variety of samples types. Although newer protein assay methods provide greater speed and convenience, the Lowry method remains a popular, accurate, and useful option for many applications.

Highlights:

  • Popular method – widely cited in protein research literature

  • Colorimetric – measure with a standard spectrophotometer or plate reader (750nm)

  • Stable – uses a modified cupric sulfate-tartrate reagent that is stable at room temperature

  • Large range – exhibits good linearity in the range 1 to 1500μg/mL (tested with BSA protein)

  • Convenient – microplate and cuvet protocols provided with the instructions

Includes:

Modified Lowry Protein Assay Reagent, Folin-Ciocalteu Phenol Reagent (2N), and ampules of Albumin Standard (2mg/mL)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sufficient For: 480 tube assays or 2400 microplate assays

• Modified Lowry Protein Assay Reagent, 480 mL
• 2N Folin-Ciocalteu Phenol Reagent, 50 mL
• Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL

Store at 4°C.
Colorimetric
Protein Quantitation Assay
480 Tube Assays or 2400 Microplate Assays
Spectrophotometer, Microplate Reader
Lowry Assay
Pierce™
Not Target-Specific
Solution-based Detection, Absorbance
530 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce Modified Lowry Protein Assay Kit

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P234-Keep only in original container.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P305-IF IN EYES:.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : sodium carbonate, Sulfuric acid copper (2+) salt (1:1), hydrate(1:5), lithium sulphate, phosphoric a

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.