Learn More
Thermo Scientific™ Pierce™ Monomeric Avidin Agarose Kit, 2 mL
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Monomeric Avidin Resins and Kit enable gentle, reversible-binding and purification of biotinylated proteins.
With typical avidin or streptavidin, the biotin-binding affinity (Kd = 10-15M) is so great that purification with these traditional media require denaturing conditions for elution, such as 8M Guanidine·HCl at pH 1.5 or boiling in reducing SDS-PAGE sample loading buffer. In addition to their adverse effects on the biotinylated protein of interest, such harsh elution conditions typically cause extraneous proteins (i.e., the avidin or streptavidin subunits) to strip from the resin and co-elute with the desired purification product. By contrast, when avidin is coupled to a resin (e.g., crosslinked beaded agarose or UltraLink Resin) as the subunit monomer, its specificity for biotin is retained, but its biotin-binding affinity decreases to a level (Kd = 10-8M ) that is conducive to subsequent elution.
Highlights:
Non-denaturing – purifies biotinylated products using mild elution conditions (2mM free biotin)
Reusable – Monomeric Avidin Agarose can be regenerated and reused at least 10 times, with only a marginal loss in biotin binding capacity (approximately 2.5% decrease per regeneration)
Specific – retains biotin-binding specificity and low nonspecific binding of native avidin
Flexible – bind samples in a variety of physiological buffers and elute either with 0.1M glycine or by competition with 2mM biotin
Good binding capacity – greater than 1.2mg biotinylated BSA/mL of resin
Convenient – purchase standalone agarose or UltraLink™ Resins or a complete purification kit with prepacked column and buffers
Item-Specific Features:
- Agarose Resins and Kit are based on crosslinked 4% beaded agarose
- UltraLink Resin is an acrylamide support (UltraLink Biosupport)
Includes:
Kit contains a pre-packed column of Monomeric Avidin Agarose and binding and elution buffers.
Specifications
Specifications
| Capacity (Metric) | 2 mL |
| Description | Pierce Monomeric Avidin Agarose Kit, 2mL |
| Content And Storage | Sufficient For: Binding ≥2.4mg biotinylated BSA per column • Pierce Monomeric Avidin Column, 2 mL, 1 column • PBS Pack (makes 500 mL), 1 pack • Biotin Binding and Elution Buffers, 200 mL • Regeneration Buffer, 250 mL • Column Accessories Store kit at 4°C. |
| Product Line | Pierce |
| Quantity | 1 Column Kit |
| Target | Biotin-Tagged Molecule |
| Type | Agarose |
| Stationary Phase | Avidin |
| Format | Gravity Flow Column |
| Column Type | Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.