missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Monomeric Avidin Agarose

Reversibly bind biotinylated proteins and other ligands with monomers of avidin covalently attached as individual subunits onto beaded resins.

626.00 € - 1140.00 €

Specifications

Capacity (Metric) ≥1.2 mg Biotinylated BSA Binding per mL Resin
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature.
Product Line Pierce™
Target Biotin-Tagged Molecule
Stationary Phase Avidin
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10638464
View accessory products View Documents
Thermo Scientific™
20228
5 mL
626.00 €
5mL
Please to purchase this item. Need a web account? Register with us today!
11866744
View accessory products View Documents
Thermo Scientific™
20267
10 mL
1140.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific™ Pierce Monomeric Avidin Resins and Kit enable gentle, reversible-binding and purification of biotinylated proteins.

Pierce Monomeric Avidin Kit and Resins are unique among biotin-binding supports in allowing non-denaturing affinity purification of biotinylated molecules. The immobilized monomeric avidin protein binds biotin with high specificity and moderate affinity, allowing non-biotin molecules to be washed away and then the bound biotin-labeled molecules to be competitively eluted using 2mM biotin in phosphate buffered saline (PBS). This technique provides the gentlest elution conditions for biotinylated protein purification and avoids the contamination and other problems associated with traditional avidin and streptavidin methods. The monomeric avidin column can then be regenerated by using 0.1M glycine to strip the column of residual bound biotin without losing the ability to bind another biotinylated sample.

With typical avidin or streptavidin, the biotin-binding affinity (Kd = 10-15M) is so great that purification with these traditional media require denaturing conditions for elution, such as 8M Guanidine·HCl at pH 1.5 or boiling in reducing SDS-PAGE sample loading buffer. In addition to their adverse effects on the biotinylated protein of interest, such harsh elution conditions typically cause extraneous proteins (i.e., the avidin or streptavidin subunits) to strip from the resin and co-elute with the desired purification product. By contrast, when avidin is coupled to a resin (e.g., crosslinked beaded agarose or UltraLink Resin) as the subunit monomer, its specificity for biotin is retained, but its biotin-binding affinity decreases to a level (Kd = 10-8M ) that is conducive to subsequent elution.

Highlights:

  • Non-denaturing – purifies biotinylated products using mild elution conditions (2mM free biotin)

  • Reusable – Monomeric Avidin Agarose can be regenerated and reused at least 10 times, with only a marginal loss in biotin binding capacity (approximately 2.5% decrease per regeneration)

  • Specific – retains biotin-binding specificity and low nonspecific binding of native avidin

  • Flexible – bind samples in a variety of physiological buffers and elute either with 0.1M glycine or by competition with 2mM biotin

  • Good binding capacity – greater than 1.2mg biotinylated BSA/mL of resin

  • Convenient – purchase standalone agarose or UltraLink™ Resins or a complete purification kit with prepacked column and buffers

Item-Specific Features:

  • Agarose Resins and Kit are based on crosslinked 4% beaded agarose
  • UltraLink Resin is an acrylamide support (UltraLink Biosupport)

Includes:

Kit contains a pre-packed column of Monomeric Avidin Agarose and binding and elution buffers.

Specifications

Specifications

≥1.2 mg Biotinylated BSA Binding per mL Resin
Pierce™
Avidin
Affinity, Agarose Resin
Upon receipt store product at 4°C. Product is shipped at ambient temperature.
Biotin-Tagged Molecule
Bottle
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.