Learn More
Thermo Scientific™ BCIP Substrate Powder (5-bromo-4-chloro-3'-indolylphosphate p-toluidine)
Powders and ready-made nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolylphosphate solutions for chromogenic detection of alkaline phosphatase probes.
Brand:  Thermo Scientific™ 34040
Product Code. 10589463
  • 362.00 € / 1g
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolylphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.
The combination of NBT (nitro-blue tetrazolium chloride) and BCIP (5-bromo-4-chloro-3'-indolylphosphate p-toluidine salt) yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, a popular enzyme conjugate for antibody probes. Pierce NBT and BCIP powders are available individually and in two convenient, ready-to-use solutions (with and without a levamisole suppressor of endogenous phosphatase activity).
Highlights:
  • AP substrate – for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples
  • Chromogenic – no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph
  • Package options – choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor
1-Step™ NBT/BCIP
  • Ideal for immunoblotting applications
  • Minimal assay-to-assay variability
1-Step NBT/BCIP Plus Suppressor
  • Ideal for staining tissues assayed with calf intestinal alkaline phosphatase without the interference caused by endogenous phosphatases
  • Contains 1mM levamisole to inhibit endogenous phosphatase activity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pierce BCIP Substrate
Colorimetric
Nitrocellulose, PVDF
1 g
Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature.
Substrate
Powder
Pierce™
Alkaline Phosphatase