missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step™ NBT/BCIP plus Suppressor Substrate Solution

Powders and ready-made nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate solutions for chromogenic detection of alkaline phosphatase probes.

Brand:  Thermo Scientific™ 34070

Product Code. 11819280

  • 159.00 € / 100mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.

The combination of NBT (nitro-blue tetrazolium chloride) and BCIP (5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt) yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, a popular enzyme conjugate for antibody probes. Pierce NBT and BCIP powders are available individually and in two convenient, ready-to-use solutions (with and without a levamisole suppressor of endogenous phosphatase activity).

Highlights:

  • AP substrate – for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples

  • Chromogenic – no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph

  • Package options – choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor

1-Step™ NBT/BCIP

  • Ideal for immunoblotting applications
  • Minimal assay-to-assay variability

1-Step NBT/BCIP Plus Suppressor

  • Ideal for staining tissues assayed with calf intestinal alkaline phosphatase without the interference caused by endogenous phosphatases
  • Contains 1mM levamisole to inhibit endogenous phosphatase activity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pierce 1-Step NBT/BCIP Plus Suppressor Substrate Solution
Colorimetric
Nitrocellulose, PVDF
100 mL
Alkaline Phosphatase
Substrate and Suppressor
Liquid
1-Step™
Alkaline Phosphatase
Upon receipt store at 4°C. Product is shipped with an ice pack.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.