missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ NeutrAvidin™ Agarose

Acrylamide and agarose beads and columns of specially-modified, pH-stabilized avidin; the low-cost alternative to streptavidin biotin-binding supports.

418.00 € - 590.00 €

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze resin. Product is shipped at ambient temperature.
Particle Size 45 to 165 μm
Product Line NeutrAvidin™, Pierce™
Target Biotin-Tagged Molecule
Type Agarose
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11885835
View Documents
Thermo Scientific™
29201
10 mL
590.00 €
10mL
In Stock
to see stock.
Add to basket
 
10424005
View Documents
Thermo Scientific™
29200
5 mL
418.00 €
5mL
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Thermo Scientific™ Pierce NeutrAvidin Resins provide less nonspecific binding and cleaner results than avidin or streptavidin for many kinds of biotin affinity procedures.

Additional Information and Online Ordering

NeutrAvidin Agarose and UltraLink™ Resins are superior alternatives to avidin and streptavidin beads in many biotin-based affinity applications. NeutrAvidin Protein is a modified avidin derivative that combines several key features to provide a biotin-binding protein with exceptionally low nonspecific binding properties. Native avidin is heavily glycosylated, resulting in its high pI and nonspecific binding properties. By contrast NeutrAvidin Protein contains no carbohydrate and has a near-neutral isoelectric point (6.3), features that provide for very low nonspecific binding and broader utility in a variety of applications.

NeutrAvidin Resins are prepared by covalently coupling NeutrAvidin Protein to the beaded agarose or UltraLink Biosupport using efficient and stable chemistries, resulting in NeutrAvidin Agarose and UltraLink Resins that are leach-resistant and stable at pH 2-11. The products are excellent choices for a variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.

Highlights:

  • Low nonspecific binding – unlike avidin, NeutrAvidin Protein has no carbohydrates, eliminating nonspecific binding problems caused by sugars; unlike streptavidin, NeutrAvidin Protein has no Arg-Tyr-Asp sequence, which can be a source of binding to cell surface molecules

  • Versatile resins – agarose (polysaccharide-based) and UltraLink Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods

  • Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)

  • Superior performance – High Capacity Resin has higher binding capacity than best alternative supplier streptavidin products, allowing use of small amounts of resin for experiments

Item-Specific Features:

  • Agarose Resins are crosslinked 6% beaded agarose
  • UltraLink Resins are beaded acrylamide support (UltraLink Biosupport)
  • Plus and High Capacity Resins have a higher loading of NeutrAvidin Protein than the regular forms

Recommended for:

Immunoprecipitation; Purifying proteins that bind to biotinylated ligands; Capturing biotinylated cell-surface proteins; Purifying biotinylated peptides

Specifications

Specifications

Upon receipt store at 4°C. Do not freeze resin. Product is shipped at ambient temperature.
NeutrAvidin™, Pierce™
Agarose
Bottle
45 to 165 μm
Biotin-Tagged Molecule
NeutrAvidin Protein
Affinity, Agarose Resin
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.