missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ NeutrAvidin™ Coated High Capacity Plates

Product Code. 10791275 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Color:
Black
Clear
White
No. of Wells:
96
384
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence
missing translation for 'unitSize'
Pack of 5
This item is not returnable. View return policy

Product Code. 10791275

missing translation for 'mfr': Thermo Scientific™ 15507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Pierce NeutrAvidin High Binding Capacity (HBC) Coated Plates are pre-blocked and ready-to-use polystyrene microplates for assays involving biotinylated antibodies or biotin-labeled molecules.

Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. With a near-neutral pI (6.3), this results in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein lacks the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells.

Thermo Scientific NeutrAvidin™ Coated High Capacity Plates are designed for binding biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Features of Thermo Scientific Pierce Coated Plates include:

  • Unique plate-coating technology – results in high loading and binding capacity
  • NeutrAvidin Protein – coated plates have up to twice the binding capacity of typical plates
  • Robust and sensitive – NeutrAvidin Protein yields less nonspecific binding for improved signal-to-noise ratios
  • Broader dynamic range – extends the quantitative range so there is less need for dilutions
  • Save time – pre-blocked plates to reduce the number of assay steps
  • Flexible assay formats – choose from 96- and 384-well formats in a variety of colors

The proprietary plate-coating technology used to manufacture these Thermo Scientific Pierce Coated Plates yields high-capacity plates with a wider detection limit than regular binding capacity plates. The standard curve exhibits greater linearity for detecting small biotinylated molecules such as peptides and oligonucleotides, resulting in greater assay precision.

For detection of low concentrations (ng/mL) of biotinylated antibodies and other proteins, check out

Pierce™ NeutrAvidin™ Coated High Sensitivity Plates, Clear, 8-Well Strip

TRUSTED_SUSTAINABILITY

Specifications

Binding Property High Capacity, Affinity
Format Standard
Content And Storage Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C.
Detection Method Colorimetric
Product Line NeutrAvidin Pierce
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment NeutrAvidin Protein
No. per Pack 5 Plates
Plate Blocking SuperBlock™ Buffer
Quantity 5 plates
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.