missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NeutrAvidin Protein, HRP

NeutrAvidin Protein, HRP conjugate, 31001, from Invitrogen™, 31001, from Invitrogen™ Species Reactivity: ; Applications: ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.

Brand:  Thermo Scientific™ 31001

332.91 EUR valid until 2024-12-31
Use promo code "21979" to get your promotional price.


Product Code. 11825855

This item is not returnable. View return policy

Description

Description

NeutrAvidin Protein, HRP conjugate, 31001, from Invitrogen., 31001, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.

NeutrAvidin Protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. Purified and conjugated forms of NeutrAvidin Protein provide exceptional performance in Western blot, ELISA and IHC applications that require biotin-binding probes. Assay specificity, sensitivity, and signal-to-noise ratios with NeutrAvidin Protein are generally equivalent or better than with the significantly more expensive streptavidin. Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14µg/mg of protein, which is near the theoretical maximum activity.
Formulation: 2mg purified protein and no preservative
31001 contains 2 mg of lyophilized powder from modified avidin in salt-free solution.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HRP
Upon receipt store product 4°C. Product is shipped at ambient temperature.
Avidin
HRP (Horseradish Peroxidase)
2 mg
Solid
Biotin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.