Learn More
Thermo Scientific™ Pierce™ Ninhydrin (2,2-dihydroxy-1H-indene-1,3(2H)-dione)
Label, detect and measure peptides, proteins and amino acids by HPLC (liquid chromatography) using this amine-reactive, colored reagent (Amax 440nm).
Brand:  Thermo Scientific™ 21003BID
Product Code. 11876794
  • 1356.00 € / 500g
Explore more special offers
Description
Description
Thermo Scientific™ Pierce Ninhydrin is a classic, amine-reactive, colorimetric detection reagent for protein detection and amino acid analysis.
Highlights:
  • Used in amino acid chromatography
  • Offers superb color response and low blank
Ordering Alerts:
Indefinitely stable. No refrigeration required. Keep bottle tightly sealed. Avoid exposure to direct sunlight and ammonia.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ninhydrin
Pierce™
Other Label or Dye
500 g
Amine
Store in a segregated and approved area. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Colorimetric
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Pierce™ Ninhydrin (2,2-dihydroxy-1H-indene-1,3(2H)-dione) > 500g