Learn More
Thermo Scientific™ PITC (Edman's Reagent) (phenylisothiocyanate)
Phenylisothiocyanate, an amine-reactive, colored compound (Amax 254nm; UV) for pre-column derivatization of amino acids for reverse-phase HPLC.
Brand: Thermo Scientific™ 26922
168.15 EUR valid until 2025-03-14
Use promo code "24142" to get your promotional price.
Description
Thermo Scientific™ Pierce PITC (phenylisothiocyanate) is a high-purity reagent for pre-column quantitative derivatization of amino acids by reverse-phase HPLC.
Highlights:
- Also known as Edman's Reagent
- Reacts readily with amino acids in 5 to 10 minutes at room temperature
- Resulting phenylthiocarbamyl derivatives can be separated and quantified in 30 minutes using reverse-phase HPLC to produce stable products with all amino acids including proline
Ordering Alerts:
X in the ordering table indicates that hazardous shipping charges apply.
Specifications
PITC | |
Upon receipt store at room temperature. | |
10 x 1 mL |
Amine | |
Other Label or Dye | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.